Learn More
2-(Methylamino)octanoic Acid, TRC
Product Code. 30476419
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30476419 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30476419 Supplier Toronto Research Chemicals Supplier No. TRCM28705310MG
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-(Methylamino)octanoic Acid
CAS 101080-67-5
Molecular Formula C9H19NO2
Quantity 10 mg
InChI Formula InChI=1S/C9H19NO2/c1-3-4-5-6-7-8(10-2)9(11)12/h8,10H,3-7H2,1-2H3,(H,11,12)
SMILES CCCCCCC(C(=O)O)NC
IUPAC Name 2-(methylamino)octanoic acid
Molecular Weight (g/mol) 173.25
Formula Weight 173.1416