Learn More
2-Methyl Ethynyl Estradiol, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Methyl Ethynyl Estradiol |
| CAS | 3240-39-9 |
| Molecular Formula | C21 H26 O2 |
| Quantity | 5 mg |
| Synonym | (17α)-2-Methyl-19-norpregna-1,3,5(10)-trien-20-yne-3,17-diol, 19-Nor-17α-pregna-1,3,5(10)-trien-20-yne-3,17-diol','2-methyl- (7CI), 2-Methyl-19-nor-17α-pregna-1,3,5(10)-trien-20-yne-3,17-diol (2-Methyl-ethinylestradiol), 2-Methyl Ethynyl Estradiol, Ethinylestradiol Imp M (EP) |
| InChI Formula | InChI=1S/C21H26O2/c1-4-21(23)10-8-18-16-6-5-14-12-19(22)13(2)11-17(14)15(16)7-9-20(18,21)3/h1,11-12,15-16,18,22-23H,5-10H2,2-3H3/t15-,16+,18-,20-,21-/m0/s1 |
| SMILES | Cc1cc2[C@H]3CC[C@@]4(C)[C@@H](CC[C@@]4(O)C#C)[C@@H]3CCc2cc1O |
| IUPAC Name | (8R,9S,13S,14S,17R)-17-ethynyl-2,13-dimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthrene-3,17-diol |
| Molecular Weight (g/mol) | 310.43 |
| Formula Weight | 310.1933 |