Learn More
(±)-2-Methyl-d3-butyric Acid, CDN
Specifications
Specifications
| Chemical Name or Material | 2-(trideuteriomethyl)butanoic acid |
| CAS | 95926-92-4 |
| Molecular Formula | C5 2H3 H7 O2 |
| Quantity | 0.5 g |
| InChI Formula | InChI=1 S/C5H10O2/c1-3-4(2)5(6)7/h4H,3H2,1-2H3,(H,6,7)/i2D3 |
| SMILES | [2 H]C([2 H])([2 H])C(CC)C(=O)O |
| IUPAC Name | 2-(trideuteriomethyl)butanoic acid |
| Molecular Weight (g/mol) | 105.15 |
| Formula Weight | 105.087 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |