Learn More
2-Methyl-9H-carbazole, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Methyl-9H-carbazole |
| CAS | 3652-91-3 |
| Molecular Formula | C13H10N |
| Quantity | 2 mg |
| InChI Formula | InChI=1S/C13H11N/c1-9-6-7-11-10-4-2-3-5-12(10)14-13(11)8-9/h2-8,14H,1H3 |
| SMILES | Cc1ccc2c(c1)[nH]c3ccccc23 |
| IUPAC Name | 2-methyl-9H-carbazole |
| Molecular Weight (g/mol) | 181.23 |
| Formula Weight | 181.0891 |