missing translation for 'onlineSavingsMsg'
Learn More

2'-Methyl-5'-(trifluoromethyl)acetophenone, 97%, Thermo Scientific™

Product Code. 15474457
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15474457 1 g 1g
15484457 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 15474457

Brand: Thermo Scientific Alfa Aesar H31819.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 725743-50-0
Molecular Formula C10H9F3O
Molecular Weight (g/mol) 202.18
MDL Number MFCD09832290
InChI Key ZLDDSCHHMXTRHJ-UHFFFAOYSA-N
Synonym 2'-methyl-5'-trifluoromethyl acetophenone, 1-2-methyl-5-trifluoromethyl-phenyl-ethanone, 1-2-methyl-5-trifluoromethyl phenyl ethan-1-one, ethanone, 1-2-methyl-5-trifluoromethyl phenyl, 1-2-methyl-5-trifluoromethyl phenyl ethanone, 1-acetyl-2-methyl-5-trifluoromethyl benzene
PubChem CID 46737522
SMILES CC(=O)C1=C(C)C=CC(=C1)C(F)(F)F

Specifications

Quantity 1 g
IUPAC Name 1-[2-methyl-5-(trifluoromethyl)phenyl]ethan-1-one
Formula Weight 202.17
Percent Purity 97%
Chemical Name or Material 2'-Methyl-5'-(trifluoromethyl)acetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.