Learn More
2-Methyl-2-thiopropylamine Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Methyl-2-thiopropylamine Hydrochloride |
| CAS | 32047-53-3 |
| Molecular Formula | C4H12ClNS |
| Quantity | 50 mg |
| Synonym | 1-Amino-2-methyl-2-propanethiol Hydrochloride, 2-Mercapto-2-methyl-1-propylamine Hydrochloride, 2-Methyl-3-aminopropane-2-thiol Hydrochloride |
| InChI Formula | InChI=1S/C4H11NS.ClH/c1-4(2,6)3-5;/h6H,3,5H2,1-2H3;1H |
| SMILES | Cl.CC(C)(S)CN |
| IUPAC Name | 1-amino-2-methylpropane-2-thiol;hydrochloride |
| Molecular Weight (g/mol) | 141.66 |
| Formula Weight | 141.0379 |