missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-pentanol, 99%

Product Code. 11409396
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409396 100 g 100g
11489386 5 g 5g
11499386 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409396 Supplier Thermo Scientific Alfa Aesar Supplier No. A19090.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 590-36-3 | C6H14O | 102.18 g/mol

It is used to study its antagonist activity. It reacts with a second target site to alter the spatial relation between the hydrophobic agonist binding site and the allosteric site. It may initiate the polymerization of isocyanates and epoxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to study its antagonist activity. It reacts with a second target site to alter the spatial relation between the hydrophobic agonist binding site and the allosteric site. It may initiate the polymerization of isocyanates and epoxides.

Solubility
Soluble in water at 20°C 32g/L. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-36-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00004485
InChI Key WFRBDWRZVBPBDO-UHFFFAOYSA-N
Synonym 2-methyl-2-pentanol, 2-pentanol, 2-methyl, 2-hydroxy-2-methylpentane, unii-hu2so831kp, dimethylpropylcarbinol, hu2so831kp, 1,1-dimethylbutanol, methyl-2 pentanol-2, 2-methyl-2-hydroxypentane, acmc-209m9l
PubChem CID 11543
IUPAC Name 2-methylpentan-2-ol
SMILES CCCC(C)(C)O

Specifications

Melting Point -103°C
Density 0.81
Boiling Point 120°C to 122°C
Flash Point 30°C (86°F)
Odor Alcohol-like
Refractive Index 1.41
Quantity 100 g
UN Number UN2560
Beilstein 1718951
Solubility Information Soluble in water at 20°C 32g/L. Soluble in alcohol.
Formula Weight 102.18
Percent Purity 99%
Chemical Name or Material 2-Methyl-2-pentanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.