Learn More
2-Methyl-2-(methylamino)propan-1-ol (~85%), TRC
Specifications
Specifications
| Chemical Name or Material | 2-Methyl-2-(methylamino)propan-1-ol |
| CAS | 27646-80-6 |
| Molecular Formula | C5H13NO |
| Quantity | 1 g |
| Synonym | 2-Methyl-2-(methylamino)-1-propanol, 2-Methyl-2-methylaminopropan-1-ol, 2-Methylamino-2-methyl-1-propanol, N-(2-Hydroxy-1,1-dimethylethyl)-N-methylamine, NSC 147612 |
| InChI Formula | InChI=1S/C5H13NO/c1-5(2,4-7)6-3/h6-7H,4H2,1-3H3 |
| SMILES | CNC(C)(C)CO |
| IUPAC Name | 2-methyl-2-(methylamino)propan-1-ol |
| Molecular Weight (g/mol) | 103.16 |
| Formula Weight | 103.0997 |