missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-imidazoline, 95%, Thermo Scientific Chemicals

Product Code. 11976271 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11976271 25 g 25g
11966271 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11976271 Supplier Thermo Scientific Acros Supplier No. 439720250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 534-26-9
Molecular Formula C4H8N2
Molecular Weight (g/mol) 84.12
InChI Key VWSLLSXLURJCDF-UHFFFAOYSA-N
Synonym 2-methyl-2-imidazoline, 2-methylimidazoline, 1h-imidazole, 4,5-dihydro-2-methyl, methylglyoxalidine, 2-imidazoline, 2-methyl, 4,5-dihydro-2-methyl-1h-imidazole, unii-987f50e3pe, 2-methyl imidazoline, 2-methyl-4,5-dihydroimidazole, 2-methyl-20imidazolin
PubChem CID 10798
IUPAC Name 2-methyl-4,5-dihydro-1H-imidazole
SMILES CC1=NCCN1

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 104°C to 107°C
Color Colorless to Yellow or White to Yellow
Boiling Point 110°C (2.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 95%
Quantity 25 g
Merck Index 15,5696
Formula Weight 84.12
Percent Purity 95%
Physical Form Crystals, Powder or Needles
Chemical Name or Material 2-Methyl-2-imidazoline
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methyl-2-imidazoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.