Learn More
(±)-2-Methyl-2,4-pentane-d12-diol, CDN
Specifications
Specifications
| Chemical Name or Material | 2-Methyl-2,4-pentanediol-D12 |
| CAS | 284474-72-2 |
| Molecular Formula | C6 D12 H2 O2 |
| Quantity | 0.5 g |
| Synonym | 2,4-Pentane-1,1,1,2,3,3,5,5,5-d9-diol, 4-(methyl-d3)- (9 CI), (±)-2-Methyl-2,4-pentane-d12-diol, 4-(Methyl-d3)-2,4-pentane-1,1,1,2,3,3,5,5,5-d9-diol, 2-Methyl-2,4-pentanediol-d12, 2-Methyl-2,4-pentanediol-D12, Hexylene Glycol-d12 |
| InChI Formula | InChI=1 S/C6H14O2/c1-5(7)4-6(2,3)8/h5,7-8 H,4H2,1-3H3/i1D3,2D3,3D3,4D2,5 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(O)C([2 H])([2 H])C(O)(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1,1,2,3,3,5,5,5-nonadeuterio-4-(trideuteriomethyl)pentane-2,4-diol |
| Molecular Weight (g/mol) | 130.2481 |
| Formula Weight | 130.1747 g/mol |
| Show More |