Learn More
2-Methoxy-d3-pyrazine, CDN
Product Code. 30327258
Change view
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30327258 0.05 g 0.05g
30314997 0.1 g 0.10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30327258 Supplier CDN Supplier No. CDND73240.05G
Please to purchase this item. Need a web account? Register with us today!
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Methoxypyrazine-D3 (methoxy-D3)
CAS 32046-21-2
Molecular Formula C5 D3 H3 N2 O
Quantity 0.05 g
Synonym Pyrazine, methoxy-d3- (8 CI), 2-Methoxy-d3-pyrazine, 2-Methoxypyrazine-D3 (methoxy-D3), Methoxy-d3-pyrazine
InChI Formula InChI=1 S/C5H6N2O/c1-8-5-4-6-2-3-7-5/h2-4 H,1H3/i1D3
SMILES [2 H]C([2 H])([2 H])Oc1cnccn1
IUPAC Name 2-(trideuteriomethoxy)pyrazine
Molecular Weight (g/mol) 113.1324
Formula Weight 113.0668 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity