Learn More
2-Methoxy-4-(2-nitro-1-propenyl)phenol, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Methoxy-4-(2-nitro-1-propenyl)phenol |
| CAS | 5395-47-1 |
| Molecular Formula | C10H11NO4 |
| Quantity | 250 mg |
| Synonym | 2-Methoxy-4-(2-nitropropenyl)phenol, NSC 3298, NSC 57759 |
| InChI Formula | InChI=1S/C10H11NO4/c1-7(11(13)14)5-8-3-4-9(12)10(6-8)15-2/h3-6,12H,1-2H3/b7-5+ |
| SMILES | COc1cc(\C=C(/C)\[N+](=O)[O-])ccc1O |
| IUPAC Name | 2-methoxy-4-[(E)-2-nitroprop-1-enyl]phenol |
| Molecular Weight (g/mol) | 209.2 |
| Formula Weight | 209.0688 |