Learn More
2-Methoxy-3-(trifluoromethyl)phenylacetic acid, 97%, Thermo Scientific™
Product Code. 15467865
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15467865 250 mg 250mg
15457865 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15467865 Supplier Thermo Scientific Alfa Aesar Supplier No. H31671.MD
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1017778-84-5
Molecular Formula C10H9F3O3
Molecular Weight (g/mol) 234.17
MDL Number MFCD09832317
InChI Key LMQFQFJTFHHNQH-UHFFFAOYSA-N
Synonym 2-methoxy-3-trifluoromethyl phenylacetic acid, 2-2-methoxy-3-trifluoromethyl phenyl acetic acid, 2-methoxy-3-trifluoromethyl phenyl acetic acid, 2-methoxy-3-trifluoromethyl phenylacetic acid, jrd
PubChem CID 46737592
IUPAC Name 2-[2-methoxy-3-(trifluoromethyl)phenyl]acetic acid
SMILES COC1=C(CC(O)=O)C=CC=C1C(F)(F)F
Specifications
Melting Point 71°C to 75°C
Quantity 250 mg
Formula Weight 234.18
Percent Purity 97%
Chemical Name or Material 2-Methoxy-3-(trifluoromethyl)phenylacetic acid