Learn More
2-Mercaptoethanol-1,1,2,2-d4, CDN
Specifications
Specifications
| Chemical Name or Material | 2-Mercaptoethanol-D4 |
| CAS | 284474-53-9 |
| Molecular Formula | C2 D4 H2 O S |
| Quantity | 0.25 g |
| Synonym | Ethan-1,1,2,2-d4-ol, 2-mercapto- (9 CI), 2-Mercaptoethanol-1,1,2,2-d4, 2-Mercaptoethanol-D4, 2-Mercaptoethanol-d4, 2-Mercaptoethan-1,1,2,2-d4-ol, 2-Sulfanylethanol-d4, β-Hydroxyethanethiol-d4 |
| InChI Formula | InChI=1 S/C2H6OS/c3-1-2-4/h3-4 H,1-2H2/i1D2,2D2 |
| SMILES | [2 H]C([2 H])(O)C([2 H])([2 H])S |
| IUPAC Name | 1,1,2,2-tetradeuterio-2-sulfanylethanol |
| Molecular Weight (g/mol) | 82.1581 |
| Formula Weight | 82.039 g/mol |
| Show More |