Learn More
2-Isopropyl-6-methyl-4-pyrimidone, TRC
Product Code. 30402996
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30402996 5 g
30388857 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30402996 Supplier Toronto Research Chemicals Supplier No. TRCI8727205G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2814-20-2
Molecular Formula C8 H12 N2 O
Molecular Weight (g/mol) 152.19
Synonym 4(3H)-Pyrimidinone, 6-methyl-2-(1-methylethyl)- (ACI), 4(1H)-Pyrimidinone, 2-isopropyl-6-methyl- (8CI), 4(1H)-Pyrimidinone, 6-methyl-2-(1-methylethyl)- (9CI), 4-Pyrimidinol, 2-isopropyl-6-methyl- (6CI, 7CI), 6-Methyl-2-(1-methylethyl)-4(3H)-pyrimidinone (ACI), 2-Isopropyl-4-hydroxy-6-methylpyrimidine, 2-Isopropyl-4-methyl-6-hydroxypyrimidine, 2-Isopropyl-4-methyl-6-pyrimidinone, 2-Isopropyl-4-methylpyrimidin-6-ol, 2-Isopropyl-6-methyl-4(3H)-pyrimidone, 2-Isopropyl-6-methyl-4-hydroxypyrimidine, 2-Isopropyl-6-methyl-4-pyrimidinol, 2-Isopropyl-6-methyl-4-pyrimidone, 4-Hydroxy-2-isopropyl-6-methylpyrimidine, G 27550, IMHP, Pyrimidinol
IUPAC Name 6-methyl-2-propan-2-ylpyrimidin-4-ol
SMILES CC(C)c1nc(C)cc(O)n1
Specifications
Quantity 5 g
InChI Formula InChI=1S/C8H12N2O/c1-5(2)8-9-6(3)4-7(11)10-8/h4-5H,1-3H3,(H,9,10,11)
Formula Weight 152.095
Chemical Name or Material 2-Isopropyl-6-methyl-4-pyrimidinol