Learn More
2-Iodotoluene, 98%
Product Code. p-7026612
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Product Code. 11495337
Brand: Thermo Scientific Alfa Aesar B22697.14
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 615-37-2 | C7H7I | 218.037 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 615-37-2
Molecular Formula C7H7I
Molecular Weight (g/mol) 218.037
MDL Number MFCD00001042
InChI Key RINOYHWVBUKAQE-UHFFFAOYSA-N
Synonym 2-iodotoluene, o-iodotoluene, o-tolyl iodide, benzene, 1-iodo-2-methyl, o-methyliodobenzene, toluene, o-iodo, iodotoluene, ortho-iodotoluene, 2-methyliodobenzene, unii-8ok4h85t07
PubChem CID 5128
ChEBI CHEBI:39813
IUPAC Name 1-iodo-2-methylbenzene
SMILES CC1=CC=CC=C1I
Specifications
Density 1.713
Boiling Point 210°C to 212°C
Flash Point 90°C (194°F)
Refractive Index 1.609
Quantity 25 g
Beilstein 1634294
Sensitivity Light sensitive
Formula Weight 218.04
Percent Purity 98%
Chemical Name or Material 2-Iodotoluene