Learn More
2-Iodo Adenosine 2’,3’-Acetonide, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Iodo Adenosine 2',3'-Acetonide |
| CAS | 141018-25-9 |
| Molecular Formula | C13 H16 I N5 O4 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C13H16IN5O4/c1-13(2)22-7-5(3-20)21-11(8(7)23-13)19-4-16-6-9(15)17-12(14)18-10(6)19/h4-5,7-8,11,20H,3H2,1-2H3,(H2,15,17,18)/t5-,7-,8-,11-/m1/s1 |
| SMILES | CC1(C)O[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O1)n3cnc4c(N)nc(I)nc34 |
| IUPAC Name | [(3aR,4R,6R,6aR)-4-(6-amino-2-iodopurin-9-yl)-2,2-dimethyl-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-6-yl]methanol |
| Molecular Weight (g/mol) | 433.2 |
| Formula Weight | 433.0247 |