Learn More
2-Iodo-3-methoxypyridine, 97%
Product Code. 11369875
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11369875 1 g 1g
11379875 5 g 5g
11389875 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11369875 Supplier Thermo Scientific Alfa Aesar Supplier No. L19868.03
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
CAS: 93560-55-5 | C6H6INO | 235.024 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 93560-55-5
Molecular Formula C6H6INO
Molecular Weight (g/mol) 235.024
MDL Number MFCD00234181
InChI Key NJFRZBAZMPWJKQ-UHFFFAOYSA-N
Synonym 2-iodanyl-3-methoxy-pyridine, pyridine, 2-iodo-3-methoxy, 2-iodo-3-methoxy-pyridine
PubChem CID 817173
IUPAC Name 2-iodo-3-methoxypyridine
SMILES COC1=C(N=CC=C1)I
Specifications
Melting Point 57°C to 61°C
Quantity 1 g
Beilstein 115569
Sensitivity Light sensitive
Formula Weight 235.03
Percent Purity 97%
Chemical Name or Material 2-Iodo-3-methoxypyridine