Learn More
2-Hydroxymethylene Ethisterone, TRC
Product Code. 30406368
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30406368 1 g 1g
30390105 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30406368 Supplier Toronto Research Chemicals Supplier No. TRCH9467001G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2787-02-2
Molecular Formula C22 H28 O3
Molecular Weight (g/mol) 340.46
IUPAC Name (2E,8R,9S,10R,13S,14S,17S)-17-ethynyl-17-hydroxy-2-(hydroxymethylidene)-10,13-dimethyl-1,6,7,8,9,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-3-one
SMILES C[C@]12C\C(=C/O)\C(=O)C=C1CC[C@@H]3[C@@H]2CC[C@@]4(C)[C@H]3CC[C@]4(O)C#C
Specifications
Quantity 1 g
InChI Formula InChI=1S/C22H28O3/c1-4-22(25)10-8-18-16-6-5-15-11-19(24)14(13-23)12-20(15,2)17(16)7-9-21(18,22)3/h1,11,13,16-18,23,25H,5-10,12H2,2-3H3/b14-13+/t16-,17+,18+,20+,21+,22-/m1/s1
Formula Weight 340.2038
Percent Purity >95%
Chemical Name or Material 2-Hydroxymethylene Ethisterone
Grade HPLC