Learn More
2-Hydroxyethyl Methacrylate, TRC
Product Code. 30010022
Change view
Click to view available options
Quantity:
500 mg
1 g
25 g
Unit Size:
1g
25g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010022 500 mg 500mg
30135305 1 g 1g
30138398 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30010022 Supplier Toronto Research Chemicals Supplier No. TRCH942230500MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 868-77-9
Formula Weight 130.063
InChI Formula InChI=1S/C6H10O3/c1-5(2)6(8)9-4-3-7/h7H,1,3-4H2,2H3
IUPAC Name 2-hydroxyethyl 2-methylprop-2-enoate
Molecular Formula C6 H10 O3
Molecular Weight (g/mol) 130.14
SMILES CC(=C)C(=O)OCCO
Synonym 2-Propenoic acid, 2-methyl-, 2-hydroxyethyl ester, Methacrylic acid, 2-hydroxyethyl ester (6CI,8CI), Methacrylic acid, ester with glycol (7CI), 2-(Methacryloyloxy)ethanol, 2-HEMA, 2-Hydroxyethyl 2-methylprop-2-enoate, 2-Hydroxyethyl methacrylate, Acryester HISS, Acryester HO, Acryester HOMA, Acryl Ester HO, Bisomer HEMA, Bisomer SR, Blemmer E, EB 109, EB 109 (monomer), EM 321, Ethylene glycol methacrylate, Ethylene glycol monomethacrylate, GE 610, Glycol methacrylate, Glycol monomethacrylate, HEMA, HEMA 90, JB 4 Plus, Kayarad 2-HEMA, Light Ester HO, Light Ester HO 250, Light Ester HO 250M, Light Ester HO 250N, M 0085, Mhoromer BM 903, Mhoromer BM 905, Monomer MG 1, NSC 24180, Rocryl 400, Visiomer HEMA 98, β-Hydroxyethyl methacrylate
Chemical Name or Material 2-Hydroxyethyl Methacrylate
Specifications
Quantity 500 mg