Learn More
2-Hydroxyethanephosphonic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Ethephon-hydroxy |
| CAS | 22987-21-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C2 H7 O4 P |
| Quantity | 25 mg |
| Synonym | Phosphonic acid, (2-hydroxyethyl)- (8CI,9CI), P-(2-Hydroxyethyl)phosphonic acid, (2-Hydroxyethyl)phosphonic acid, 2-Hydroxyethanephosphonic acid |
| InChI Formula | InChI=1S/C2H7O4P/c3-1-2-7(4,5)6/h3H,1-2H2,(H2,4,5,6) |
| SMILES | OCCP(=O)(O)O |
| IUPAC Name | 2-hydroxyethylphosphonic acid |
| Molecular Weight (g/mol) | 126.05 |
| Show More |