Learn More
2-Hydroxybenzoic Acid-d6, CDN
Specifications
Specifications
| Chemical Name or Material | Salicylic Acid-D6 |
| CAS | 285979-87-5 |
| Molecular Formula | C7 D6 O3 |
| Quantity | 0.1 g |
| Synonym | Benzoic-2,3,4,5-d4 acid-d, 6-(hydroxy-d)- (9 CI), 2-Carboxyphenol-d6, Perdeutero-2-hydroxybenzoic acid, 2-Hydroxybenzoic Acid-d6 |
| InChI Formula | InChI=1 S/C7H6O3/c8-6-4-2-1-3-5(6)7(9)10/h1-4,8 H,(H,9,10)/i1D,2 D,3 D,4 D/hD2 |
| SMILES | [2 H]OC(=O)c1c([2 H])c([2 H])c([2 H])c([2 H])c1O[2 H] |
| IUPAC Name | deuterio 2,3,4,5-tetradeuterio-6-deuteriooxybenzoate |
| Molecular Weight (g/mol) | 144.1577 |
| Formula Weight | 144.0694 g/mol |
| Show More |