Learn More
2-Hydroxy-4-oxo-6-propyl-2-cyclohexene-1-carboxylic Acid Methyl Ester, TRC
Product Code. 30404814
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30404814 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30404814 Supplier Toronto Research Chemicals Supplier No. TRCH949865100MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Hydroxy-4-oxo-6-propyl-2-cyclohexene-1-carboxylic Acid Methyl Ester
CAS 27871-91-6
Molecular Formula C11H16O4
Quantity 100 mg
Synonym Methyl 2-Hydroxy-4-oxo-6-propylcyclohex-2-enecarboxylate
InChI Formula InChI=1S/C11H16O4/c1-3-4-7-5-8(12)6-9(13)10(7)11(14)15-2/h6-7,10,13H,3-5H2,1-2H3
SMILES CCCC1CC(=O)C=C(O)C1C(=O)OC
IUPAC Name methyl 2-hydroxy-4-oxo-6-propylcyclohex-2-ene-1-carboxylate
Molecular Weight (g/mol) 212.24
Formula Weight 212.1049