Learn More
2-Hydroxy-4-oxo-6-propyl-2-cyclohexene-1-carboxylic Acid Methyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Hydroxy-4-oxo-6-propyl-2-cyclohexene-1-carboxylic Acid Methyl Ester |
| CAS | 27871-91-6 |
| Molecular Formula | C11H16O4 |
| Quantity | 500 mg |
| Synonym | Methyl 2-Hydroxy-4-oxo-6-propylcyclohex-2-enecarboxylate |
| InChI Formula | InChI=1S/C11H16O4/c1-3-4-7-5-8(12)6-9(13)10(7)11(14)15-2/h6-7,10,13H,3-5H2,1-2H3 |
| SMILES | CCCC1CC(=O)C=C(O)C1C(=O)OC |
| IUPAC Name | methyl 2-hydroxy-4-oxo-6-propylcyclohex-2-ene-1-carboxylate |
| Molecular Weight (g/mol) | 212.24 |
| Formula Weight | 212.1049 |