missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-3-iodo-5-methylpyridine, 95%, Thermo Scientific™

Product Code. p-7030201
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15473767 250 mg 250mg
11342909 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15473767 Supplier Thermo Scientific Alfa Aesar Supplier No. H27751.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as pharmaceutical intermediate.

Solubility
Soluble in dimethyl sulfoxide.

Notes
Light sensitive. Store in dark place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 313678-93-2
Molecular Formula C6H6INO
Molecular Weight (g/mol) 235.02
MDL Number MFCD09037469
InChI Key ASWXKNUHPBTHJI-UHFFFAOYSA-N
Synonym 3-iodo-5-methylpyridin-2-ol, 2-hydroxy-3-iodo-5-methylpyridine, 3-iodo-5-methylpyridin-2 1h-one, acmc-20antm, 2 1h-pyridinone,3-iodo-5-methyl
PubChem CID 42553066
IUPAC Name 3-iodo-5-methyl-1H-pyridin-2-one
SMILES CC1=CNC(=O)C(I)=C1

Specifications

Melting Point 190°C to 193°C
Quantity 250 mg
Sensitivity Light sensitive
Solubility Information Soluble in dimethyl sulfoxide.
Formula Weight 235.02
Percent Purity 95%
Chemical Name or Material 2-Hydroxy-3-iodo-5-methylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.