missing translation for 'onlineSavingsMsg'
Learn More

2-Hydrazinobenzothiazole, 97%

Product Code. p-7023945
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11459136

Brand: Thermo Scientific Alfa Aesar A18006.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 615-21-4 | C7H7N3S | 165.214 g/mol

It is employed in the spectrophotometric analysis determination of methenamine and its salts. It is also used in the detection and repair mechanism of Etheno-DNA adducts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the spectrophotometric analysis determination of methenamine and its salts. It is also used in the detection and repair mechanism of Etheno-DNA adducts.

Solubility
Solubility in hot methanol almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 615-21-4
Molecular Formula C7H7N3S
Molecular Weight (g/mol) 165.214
MDL Number MFCD00041849
InChI Key JYSUYJCLUODSLN-UHFFFAOYSA-N
Synonym 2-hydrazinobenzothiazole, 2-hydrazinylbenzo d thiazole, 2-hydrazino-1,3-benzothiazole, benzothiazolohydrazine, 2 3h-benzothiazolone, hydrazone, benzothiazol-2-ylhydrazine, benzothiazole, 2-hydrazino, 2-benzothiazolylhydrazine, usaf ek-3967, benzothiazol-2-yl-hydrazine
PubChem CID 11988
IUPAC Name 1,3-benzothiazol-2-ylhydrazine
SMILES C1=CC=C2C(=C1)N=C(S2)NN

Specifications

Melting Point 198°C to 202°C
Quantity 10 g
Formula Weight 165.21
Percent Purity 97%
Chemical Name or Material 2-Hydrazinobenzothiazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.