Learn More
2-Heptadecyn-1-ol, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Heptadecyn-1-ol |
| CAS | 2833-98-9 |
| Molecular Formula | C17H32O |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C17H32O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18/h18H,2-14,17H2,1H3 |
| SMILES | CCCCCCCCCCCCCCC#CCO |
| IUPAC Name | heptadec-2-yn-1-ol |
| Molecular Weight (g/mol) | 252.44 |
| Formula Weight | 252.2453 |