Learn More
2-Furfurylamine, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Furfurylamine |
| CAS | 617-89-0 |
| Molecular Formula | C5H7NO |
| Quantity | 100 g |
| Synonym | 2-Furanmethanamine, (2-Furanylmethyl)amine, (Furan-2-yl)methanamine, 2-(Aminomethyl)furan, 2-Furfurylamine, 2-Furylmethanamine, NSC 49136, NSC 61168 |
| InChI Formula | InChI=1S/C5H7NO/c6-4-5-2-1-3-7-5/h1-3H,4,6H2 |
| SMILES | C1=COC(=C1)CN |
| Molecular Weight (g/mol) | 97.12 |
| Formula Weight | 97.0528 |