missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoropropionic acid, 96%, Thermo Scientific Chemicals

Product Code. 15423317
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15423317 250 mg 250mg
11455570 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15423317 Supplier Thermo Scientific Alfa Aesar Supplier No. H25841.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

2-Fluoropropionic acid is used in the medical field for diagnostic purposes. It is labeled with 18F and is used as an imaging agent for detecting prostate cancer in humans. It is also used to prepare 2-fluoro fatty acids, compounds that exhibit antifungal properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoropropionic acid is used in the medical field for diagnostic purposes. It is labeled with 18F and is used as an imaging agent for detecting prostate cancer in humans. It is also used to prepare 2-fluoro fatty acids, compounds that exhibit antifungal properties.

Solubility
Partly miscible in water.

Notes
Store in a cool, dry place in tightly closed container. Store away from oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6087-13-4
Molecular Formula C3H5FO2
Molecular Weight (g/mol) 92.069
MDL Number MFCD06247714
InChI Key ZVZPFTCEXIGSHM-UHFFFAOYSA-N
Synonym 2-fluoropropionic acid, 2-fluoro-propionic acid, propanoic acid, 2-fluoro, pubchem13698, propanoic acid,2-fluoro, acmc-1b20x, ksc492i2p
PubChem CID 256876
IUPAC Name 2-fluoropropanoic acid
SMILES CC(C(=O)O)F

Specifications

Density 1.214
Boiling Point 66°C to 67°C (30 mmHg)
Flash Point 82°C (180°F)
Odor Acrid
Refractive Index 1.383
Quantity 250 mg
UN Number UN3265
Solubility Information Partly miscible in water.
Formula Weight 92.07
Percent Purity 96%
Chemical Name or Material 2-Fluoropropionic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.