Learn More
2-Fluorobenzonitrile-d4, CDN
Specifications
Specifications
| Chemical Name or Material | 2-Fluorobenzonitrile-D4 |
| CAS | 2256740-46-0 |
| Molecular Formula | C7 D4 F N |
| Quantity | 0.5 g |
| Synonym | Benzonitrile-2,3,4,5-d4, 6-fluoro- (ACI), 6-Fluorobenzonitrile-2,3,4,5-d4 (ACI), 2-Fluorobenzonitrile-d4, 2-Fluorobenzonitrile-D4 |
| InChI Formula | InChI=1 S/C7H4FN/c8-7-4-2-1-3-6(7)5-9/h1-4 H/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C#N)c(F)c1[2 H] |
| IUPAC Name | 2,3,4,5-tetradeuterio-6-fluorobenzonitrile |
| Molecular Weight (g/mol) | 125.1364 |
| Formula Weight | 125.0579 g/mol |
| Show More |