missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(trifluoromethyl)phenylboronic acid, 95+%, Thermo Scientific™

Product Code. 10778235 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10778235 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10778235 Supplier Thermo Scientific Acros Supplier No. 397090010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 352535-96-7
CAS Min % 95.0
CAS Max % 100.0
Melting Point 104°C to 109°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Molecular Formula C7H5BF4O2
Linear Formula CF3C6H3(F)B(OH)2
MDL Number MFCD03701538
Quantity 1 g
Synonym 2-fluoro-5-trifluoromethyl phenylboronic acid, 2-fluoro-5-trifluoromethyl phenyl boronic acid, 2-fluoro-5-trifluoromethyl benzeneboronic acid, 2-fluoro-5-trifluoromethyl boronic acid, 2-fluoro-5-trifluoromethylphenylboronic acid, boronic acid, 2-fluoro-5-trifluoromethyl phenyl, pubchem5041, acmc-1afry
InChI Key KUHVVLFCTMTYGR-UHFFFAOYSA-N
SMILES B(C1=C(C=CC(=C1)C(F)(F)F)F)(O)O
IUPAC Name [2-fluoro-5-(trifluoromethyl)phenyl]boronic acid
Molecular Weight (g/mol) 207.92
PubChem CID 2763301
Formula Weight 207.92
Percent Purity 95+%
Physical Form Powder
Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)phenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Fluoro-5-(trifluoromethyl)phenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.