Learn More
2-Fluoro-5-(3-fluoro-4-methylbenzylcarbamoyl)benzeneboronic acid, 97%, Thermo Scientific™
Product Code. 15400936
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15400936 250 mg 250mg
15490926 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15400936 Supplier Thermo Scientific Alfa Aesar Supplier No. H54230.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1449132-35-7
Molecular Formula C15H14BF2NO3
Molecular Weight (g/mol) 305.088
MDL Number MFCD20040159
InChI Key LZBLIQJRKZLMQM-UHFFFAOYSA-N
Synonym 2-fluoro-5-3-fluoro-4-methylbenzylcarbamoyl benzeneboronic acid, 2-fluoro-5-3-fluoro-4-methylphenyl methyl carbamoyl phenylboronic acid
PubChem CID 73995702
IUPAC Name [2-fluoro-5-[(3-fluoro-4-methylphenyl)methylcarbamoyl]phenyl]boronic acid
SMILES B(C1=C(C=CC(=C1)C(=O)NCC2=CC(=C(C=C2)C)F)F)(O)O
Specifications
Quantity 250 mg
Formula Weight 305.09
Percent Purity 97%
Chemical Name or Material 2-Fluoro-5-(3-fluoro-4-methylbenzylcarbamoyl)benzeneboronic acid