Learn More
2-Fluoro-4-methoxybenzyl alcohol, 97%
Product Code. 11340056
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11340056 1 g 1g
11330056 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11340056 Supplier Thermo Scientific Alfa Aesar Supplier No. H32118.03
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 405-09-4 | C8H9FO2 | 156.16 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 405-09-4
Molecular Formula C8H9FO2
Molecular Weight (g/mol) 156.16
MDL Number MFCD04116336
InChI Key AZYGOIQKPGPBTM-UHFFFAOYSA-N
Synonym 2-fluoro-4-methoxyphenyl methanol, 2-fluoro-4-methoxybenzyl alcohol, 2-fluoro-p-anisyl alcohol, 3-fluoro-4-hydroxymethyl anisole, benzenemethanol,2-fluoro-4-methoxy, 2-fluoro-4-methoxyphenyl methan-1-ol, benzyl alcohol, 4-methoxy-6-fluoro, pubchem7733, 2-fluoro-4-methoxybenzylalcohol, rarechem al bd 0456
PubChem CID 593638
IUPAC Name (2-fluoro-4-methoxyphenyl)methanol
SMILES COC1=CC(F)=C(CO)C=C1
Specifications
Quantity 1 g
Formula Weight 156.15
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-methoxybenzyl alcohol