Learn More
2-Ethylheptanoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Ethylheptanoic Acid |
| CAS | 3274-29-1 |
| Molecular Formula | C9H18O2 |
| Quantity | 250 mg |
| Synonym | 3-Octanecarboxylic Acid |
| InChI Formula | InChI=1S/C9H18O2/c1-3-5-6-7-8(4-2)9(10)11/h8H,3-7H2,1-2H3,(H,10,11) |
| SMILES | CCCCCC(CC)C(=O)O |
| IUPAC Name | 2-ethylheptanoic acid |
| Molecular Weight (g/mol) | 158.24 |
| Formula Weight | 158.1307 |