Learn More
2-Ethylbenzofuran, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Ethylbenzofuran |
| CAS | 3131-63-3 |
| Molecular Formula | C10H10O |
| Quantity | 10 g |
| Synonym | 2-Ethylbenzo[b]furan, 2-Ethylbenzofuran |
| InChI Formula | InChI=1S/C10H10O/c1-2-9-7-8-5-3-4-6-10(8)11-9/h3-7H,2H2,1H3 |
| SMILES | CCc1oc2ccccc2c1 |
| Molecular Weight (g/mol) | 146.19 |
| Formula Weight | 146.0732 |