Learn More
2-Ethyl-6-methylaniline, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Ethyl-6-methylaniline |
| CAS | 24549-06-2 |
| Molecular Formula | C9 H13 N |
| Quantity | 25 g |
| Synonym | Benzenamine, 2-ethyl-6-methyl-, o-Toluidine, 6-ethyl- (7CI,8CI), 2-Ethyl-6-methylbenzenamine, (2-Ethyl-6-methylphenyl)amine, 2-Ethyl-6-methylaniline, 2-Methyl-6-ethylaniline, 6-Ethyl-2-methylaniline, 6-Ethyl-o-toluidine, C 25702 |
| InChI Formula | InChI=1S/C9H13N/c1-3-8-6-4-5-7(2)9(8)10/h4-6H,3,10H2,1-2H3 |
| SMILES | CCc1cccc(C)c1N |
| IUPAC Name | 2-ethyl-6-methylaniline |
| Molecular Weight (g/mol) | 135.21 |
| Formula Weight | 135.1048 |