Learn More
2-ethyl-2-methyloctanoic acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2-ethyl-2-methyloctanoic acid |
| CAS | 31199-56-1 |
| Molecular Formula | C11H22O2 |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C11H22O2/c1-4-6-7-8-9-11(3,5-2)10(12)13/h4-9H2,1-3H3,(H,12,13) |
| SMILES | CCCCCCC(C)(CC)C(=O)O |
| IUPAC Name | 2-ethyl-2-methyloctanoic acid |
| Molecular Weight (g/mol) | 186.29 |
| Formula Weight | 186.162 |