Learn More
2-Ethyl-1,3-cyclopentanedione, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Ethyl-1,3-cyclopentanedione |
| CAS | 823-36-9 |
| Molecular Formula | C7 H10 O2 |
| Quantity | 1 g |
| Synonym | 2-Ethyl-1,3-cyclopentanedione |
| InChI Formula | InChI=1S/C7H10O2/c1-2-5-6(8)3-4-7(5)9/h5H,2-4H2,1H3 |
| SMILES | CCC1C(=O)CCC1=O |
| IUPAC Name | 2-ethylcyclopentane-1,3-dione |
| Molecular Weight (g/mol) | 126.153 |
| Formula Weight | 126.068 |