Learn More
2-Ethoxybenzoyl Chloride, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | 2-ethoxybenzoyl chloride |
| CAS | 42926-52-3 |
| Molecular Formula | C9 H9 Cl O2 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C9H9ClO2/c1-2-12-8-6-4-3-5-7(8)9(10)11/h3-6H,2H2,1H3 |
| SMILES | CCOc1ccccc1C(=O)Cl |
| IUPAC Name | 2-ethoxybenzoyl chloride |
| Molecular Weight (g/mol) | 184.62 |
| Formula Weight | 184.029 |