Learn More
2-(Dimethylamino)thioacetamide Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(Dimethylamino)thioacetamide Hydrochloride |
| CAS | 27366-72-9 |
| Molecular Formula | C4 H10 N2 S . Cl H |
| Quantity | 10 g |
| Synonym | Ethanethioamide, 2-(dimethylamino)-, hydrochloride (1:2), 2-(Dimethylamino)thioacetamide Hydrochloride, Nizatidine Imp. H (EP) as Hydrochloride |
| InChI Formula | InChI=1S/C4H10N2S.ClH/c1-6(2)3-4(5)7;/h3H2,1-2H3,(H2,5,7);1H |
| SMILES | Cl.CN(C)CC(=S)N |
| IUPAC Name | 2-(dimethylamino)ethanethioamide;hydrochloride |
| Molecular Weight (g/mol) | 154.66 |
| Formula Weight | 154.0331 |