Learn More
2-(Dimethoxymethyl)-5-(methoxymethyl)furan, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(Dimethoxymethyl)-5-(methoxymethyl)furan |
| CAS | 110339-34-9 |
| Molecular Formula | C9 H14 O4 |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C9H14O4/c1-10-6-7-4-5-8(13-7)9(11-2)12-3/h4-5,9H,6H2,1-3H3 |
| SMILES | COCc1oc(cc1)C(OC)OC |
| IUPAC Name | 2-(dimethoxymethyl)-5-(methoxymethyl)furan |
| Molecular Weight (g/mol) | 186.205 |
| Formula Weight | 186.089 |