Learn More
2-(Diethylamino)-N,N-diethylacetamide, TRC
Product Code. 30380100
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30380100 1 g 1g
30369866 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30380100 Supplier Toronto Research Chemicals Supplier No. TRCD4432001G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 27794-54-3
Molecular Formula C10H22N2O
Molecular Weight (g/mol) 186.29
Synonym N,N-Diethyl-2-(diethylamino)acetamide
SMILES CCN(CC)CC(=O)N(CC)CC
Specifications
Quantity 1 g
InChI Formula InChI=1S/C10H22N2O/c1-5-11(6-2)9-10(13)12(7-3)8-4/h5-9H2,1-4H3
Formula Weight 186.1732
Chemical Name or Material 2-(Diethylamino)-N,N-diethylacetamide