Learn More
2-Desmethylene-2-chloromethyl Ethacrynic Acid, TRC
Specifications
Specifications
| CAS | 27929-18-6 |
| Molecular Formula | C13 H13 Cl3 O4 |
| Quantity | 25 mg |
| Synonym | Ethacrynic Acid Impurity B |
| InChI Formula | InChI=1S/C13H13Cl3O4/c1-2-7(5-14)13(19)8-3-4-9(12(16)11(8)15)20-6-10(17)18/h3-4,7H,2,5-6H2,1H3,(H,17,18) |
| SMILES | CCC(CCl)C(=O)c1ccc(OCC(=O)O)c(Cl)c1Cl |
| IUPAC Name | 2-[2,3-dichloro-4-[2-(chloromethyl)butanoyl]phenoxy]acetic acid |
| Molecular Weight (g/mol) | 339.6 |
| Formula Weight | 337.9879 |
| Chemical Name or Material | 2-Desmethylene-2-chloromethyl Ethacrynic Acid |