Learn More
2’-Deoxy-3’,5’-di-O-benzoyl-2’,2’-difluorocytidine, TRC
Specifications
Specifications
| Chemical Name or Material | 2'-Deoxy-3',5'-di-O-benzoyl-2',2'-difluorocytidine |
| CAS | 134790-39-9 |
| Molecular Formula | C23 H19 F2 N3 O6 |
| Quantity | 1 g |
| Synonym | Cytidine, 2'-deoxy-2',2'-difluoro-, 3',5'-dibenzoate, 2'-Deoxy-3',5'-di-O-benzoyl-2',2'-difluorocytidine |
| InChI Formula | InChI=1S/C23H19F2N3O6/c24-23(25)18(34-20(30)15-9-5-2-6-10-15)16(13-32-19(29)14-7-3-1-4-8-14)33-21(23)28-12-11-17(26)27-22(28)31/h1-12,16,18,21H,13H2,(H2,26,27,31)/t16-,18-,21-/m1/s1 |
| SMILES | NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](COC(=O)c3ccccc3)[C@@H](OC(=O)c4ccccc4)C2(F)F |
| IUPAC Name | [(2R,3R,5R)-5-(4-amino-2-oxopyrimidin-1-yl)-3-benzoyloxy-4,4-difluorooxolan-2-yl]methyl benzoate |
| Molecular Weight (g/mol) | 471.41 |
| Formula Weight | 471.1242 |