Learn More
2-Chloronaphthalene-d7, TRC
Product Code. 30349283
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30349283 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30349283 Supplier Toronto Research Chemicals Supplier No. TRCC3737315MG
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Chloronaphthalene-d7
CAS 93951-84-9
Molecular Formula C10 2H7 Cl
Quantity 5 mg
Synonym Naphthalene-1,2,3,4,5,6,8-d7, 7-chloro- (9CI), 2-Chloronaphthalene D7, 2-Chloro-1,3,4,5,6,7,8-heptadeuterionaphthalene, 2-Chloronaphthalene-1,3,4,5,6,7,8-D7
InChI Formula InChI=1S/C10H7Cl/c11-10-6-5-8-3-1-2-4-9(8)7-10/h1-7H/i1D,2D,3D,4D,5D,6D,7D
SMILES [2H]c1c([2H])c([2H])c2c([2H])c(Cl)c([2H])c([2H])c2c1[2H]
IUPAC Name 2-chloro-1,3,4,5,6,7,8-heptadeuterionaphthalene
Molecular Weight (g/mol) 169.66
Formula Weight 169.0676