Learn More
2-(Chloromethyl)-3,5-dimethylpyridine Hydrochloride, TRC
Product Code. 30358982
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358982 1 g 1g
30357220 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30358982 Supplier Toronto Research Chemicals Supplier No. TRCC3694101G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 73590-93-9
Molecular Formula C8 H10 Cl N . Cl H
Molecular Weight (g/mol) 192.09
Synonym Pyridine, 2-(chloromethyl)-3,5-dimethyl-, hydrochloride (1:1), Pyridine, 2-(chloromethyl)-3,5-dimethyl-, hydrochloride (9CI), 2-(Chloromethyl)-3,5-dimethylpyridine hydrochloride
IUPAC Name 2-(chloromethyl)-3,5-dimethylpyridine;hydrochloride
SMILES Cl.Cc1cnc(CCl)c(C)c1
Specifications
Quantity 1 g
InChI Formula InChI=1S/C8H10ClN.ClH/c1-6-3-7(2)8(4-9)10-5-6;/h3,5H,4H2,1-2H3;1H
Formula Weight 191.0269
Chemical Name or Material 2-(Chloromethyl)-3,5-dimethylpyridine hydrochloride