Learn More
2-Chlorobenzyl-3,4,5,6-d4 Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Chlorobenzyl-3,4,5,6-d4 Chloride |
| CAS | 2732862-20-1 |
| Molecular Formula | C7 D4 H2 Cl2 |
| Quantity | 500 mg |
| Synonym | 2-Chlorobenzyl-3,4,5,6-d4 Chloride, 2-Chlorobenzyl Chloride-3,4,5,6-D4, 1-Chloro-2-(chloromethyl)benzene-3,4,5,6-d4, 5-Chloro-6-(chloromethyl)benzene-1,2,3,4-d4 |
| InChI Formula | InChI=1S/C7H6Cl2/c8-5-6-3-1-2-4-7(6)9/h1-4H,5H2/i1D,2D,3D,4D |
| SMILES | [2H]c1c([2H])c([2H])c(CCl)c(Cl)c1[2H] |
| IUPAC Name | 1-chloro-2-(chloromethyl)-3,4,5,6-tetradeuteriobenzene |
| Molecular Weight (g/mol) | 165.0532 |
| Formula Weight | 164.0098 |