missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroadenosine

Product Code. p-7042803
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15495979 250 mg 250mg
15485979 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15495979 Supplier Thermo Scientific Alfa Aesar Supplier No. J63810.MD
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 146-77-0 | C10H12ClN5O4 | 301.69 g/mol

2-Chloroadenosine is used as a selective A1-adenosine receptor agonist, and also Induces apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroadenosine is used as a selective A1-adenosine receptor agonist, and also Induces apoptosis.

Solubility
Soluble in water.

Notes
protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146-77-0
Molecular Formula C10H12ClN5O4
Molecular Weight (g/mol) 301.69
MDL Number MFCD00005734
InChI Key BIXYYZIIJIXVFW-SBTUYZBDNA-N
Synonym 2-chloroadenosine, 6-amino-2-chloropurine riboside, adenosine, 2-chloro, cl-ado, 2-chloroadenosinehemihydrate, unii-7w7zug45g8, 2-chloro adenosine, 2cl-ado, cado, 2 clado
PubChem CID 8974
IUPAC Name (2R,3R,4S,5R)-2-(6-amino-2-chloropurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
SMILES NC1=C2N=CN([C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O)C2=NC(Cl)=N1

Specifications

Color White
Quantity 250 mg
Beilstein 43957
Solubility Information Soluble in water.
Formula Weight 301.69
Physical Form Powder
Chemical Name or Material 2-Chloroadenosine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.