Learn More
2-chloro-N-(2-ethyl-6-methylphenyl)acetamide, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Ethyl-6-methyl-2-chloroacetanilide |
| CAS | 32428-71-0 |
| Molecular Formula | C11 H14 Cl N O |
| Quantity | 10 mg |
| Synonym | o-Acetotoluidide, 2-chloro-6'-ethyl- (6CI,8CI), 2-Chloro-N-(2-ethyl-6-methylphenyl)acetamide, 2-Chloro-2'-ethyl-6'-methylacetanilide, 2'-Ethyl-6'-methyl-2-chloroacetanilide, 2'-Methyl-6'-ethyl-α-chloroacetanilide, CGA 13656, CMEPA, N-(Chloroacetyl)-2-ethyl-6-methylaniline, 2-Ethyl-6-methyl-2-chloroacetanilide, Acetamide, 2-chloro-N-(2-ethyl-6-methylphenyl)- |
| InChI Formula | InChI=1S/C11H14ClNO/c1-3-9-6-4-5-8(2)11(9)13-10(14)7-12/h4-6H,3,7H2,1-2H3,(H,13,14) |
| SMILES | CCc1cccc(C)c1NC(=O)CCl |
| IUPAC Name | 2-chloro-N-(2-ethyl-6-methylphenyl)acetamide |
| Molecular Weight (g/mol) | 211.69 |
| Formula Weight | 211.0764 |