Learn More
2-Chloro-6-methylbenzoic acid, 97%
Product Code. 11334965
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11334965
Brand: Thermo Scientific Alfa Aesar L18348.06
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 21327-86-6 | C8H7ClO2 | 170.592 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 21327-86-6
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.592
MDL Number MFCD00045799
InChI Key CEFMMQYDPGCYMG-UHFFFAOYSA-N
Synonym 6-chloro-o-toluic acid, 2-chloro-6-methyl-benzoic acid, o-toluic acid, 6-chloro, 2-chloro-6-methylbenzoicacid, benzoic acid, 2-chloro-6-methyl, benzoic acid, 6-chloro-4-methyl, 6-chloro-2-methylbenzoic acid, pubchem4568, chloro-6-methylbenzoic acid, 2-m-tolyloxymethyl oxirane
PubChem CID 88870
IUPAC Name 2-chloro-6-methylbenzoic acid
SMILES CC1=C(C(=CC=C1)Cl)C(=O)O
Specifications
Melting Point 103°C to 105°C
Quantity 5 g
Beilstein 2614092
Formula Weight 170.6
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-methylbenzoic acid